Learn More
Abnova™ Human ORC1L Partial ORF (AAH11539, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004998-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The origin recognition complex (ORC) is a highly conserved six subunits protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is the largest subunit of the ORC complex. While other ORC subunits are stable throughout the cell cycle, the levels of this protein vary during the cell cycle, which has been shown to be controlled by ubiquitin-mediated proteolysis after initiation of DNA replication. This protein is found to be selectively phosphorylated during mitosis. It is also reported to interact with MYST histone acetyltransferase 2 (MyST2/HBO1), a protein involved in control of transcription silencing. [provided by RefSeq]
Sequence: MAHYPTRLKTRKTYSWVGRPLLDRKLHYQTYREMCVKTEGCSTEIHIQIGQFVLIEGDDDENPYVAKLLELFEDDSDPPPKKRARVQWFVRFCEVPACKRHLLGRKPGAQEspecificaciones
AAH11539 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAHYPTRLKTRKTYSWVGRPLLDRKLHYQTYREMCVKTEGCSTEIHIQIGQFVLIEGDDDENPYVAKLLELFEDDSDPPPKKRARVQWFVRFCEVPACKRHLLGRKPGAQ | |
RUO | |
ORC1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4998 | |
ORC1L (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSORC1/ORC1/PARC1 | |
ORC1L | |
Recombinant | |
wheat germ expression system |