missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OR52A1 Full-length ORF (Q9UKL2, 1 a.a. - 312 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00023538-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16157422

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 12-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: MSISNITVYMPSVLTLVGIPGLESVQCWIGIPFCAIYLIAMIGNSLLLSIIKSERSLHEPLYIFLGMLGATDIALASSIMPKMLGIFWFNVPEIYFDSCLLQMWFIHTLQGIESGILVAMALDRYVAICYPLRHANIFTHQLVIQIGTMVVLRAAILVAPCLVLIKCRFQFYHTTVISHSYCEHMAIVKLAAANVQVNKIYGLFVAFTVAGFDLTFITLSYIQIFITVFRLPQKEARFKAFNTCIAHICVFLQFYLLAFFSFFTHRFGSHIPPYIHILFSSIYLLVPPFLNPLVYGAKTTQIRIHVVKMFCS
Especificaciones

Especificaciones

Q9UKL2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HPFH1OR
OR52A1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
23538
OR52A1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSISNITVYMPSVLTLVGIPGLESVQCWIGIPFCAIYLIAMIGNSLLLSIIKSERSLHEPLYIFLGMLGATDIALASSIMPKMLGIFWFNVPEIYFDSCLLQMWFIHTLQGIESGILVAMALDRYVAICYPLRHANIFTHQLVIQIGTMVVLRAAILVAPCLVLIKCRFQFYHTTVISHSYCEHMAIVKLAAANVQVNKIYGLFVAFTVAGFDLTFITLSYIQIFITVFRLPQKEARFKAFNTCIAHICVFLQFYLLAFFSFFTHRFGSHIPPYIHILFSSIYLLVPPFLNPLVYGAKTTQIRIHVVKMFCS
RUO
OR52A1
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales