Learn More
Abnova™ Human OCA2 Partial ORF (AAH12097, 201 a.a. - 300 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004948-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes the human homologue of the mouse p (pink-eyed dilution) gene. The encoded protein is believed to be an integral membrane protein involved in small molecule transport, specifically tyrosine - a precursor of melanin. Mutations in this gene result in type 2 oculocutaneous albinism. [provided by RefSeq]
Sequence: GKLWQLLALSPLENYSVNLSSHVDSTLLQVDLAGALVASGPSRPGREEHIVVELTQADALGSRWRRPQQVTHNWTVYLNPRRSEHSVMSRTFEVLTRETVEspecificaciones
AAH12097 | |
Liquid | |
4948 | |
OCA2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BEY/BEY1/BEY2/BOCA/D15S12/EYCL/EYCL2/EYCL3/HCL3/P/PED/SHEP1 | |
OCA2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GKLWQLLALSPLENYSVNLSSHVDSTLLQVDLAGALVASGPSRPGREEHIVVELTQADALGSRWRRPQQVTHNWTVYLNPRRSEHSVMSRTFEVLTRETV | |
RUO | |
OCA2 | |
Wheat Germ (in vitro) | |
GST |