Learn More
Abnova™ Human OBSCN Partial ORF (XP_290923, 1551 a.a. - 1649 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084033-Q02.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The obscurin gene spans more than 150 kb, contains over 80 exons and encodes a protein of approximately 720 kDa. The encoded protein contains 68 Ig domains, 2 fibronectin domains, 1 calcium/calmodulin-binding domain, 1 RhoGEF domain with an associated PH domain, and 2 serine-threonine kinase domains. This protein belongs to the family of giant sacromeric signaling proteins that includes titin and nebulin, and may have a role in the organization of myofibrils during assembly and may mediate interactions between the sarcoplasmic reticulum and myofibrils. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: KAGMGPYSSPSEQVLLGGPSHLASEEESQGRSAQPLPSTKTFAFQTQIQRGRFSVVRQCWEKASGRALAAKIIPYHPKDKTAVLREYEALKGLRHPHLAEspecificaciones
XP_290923 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KAGMGPYSSPSEQVLLGGPSHLASEEESQGRSAQPLPSTKTFAFQTQIQRGRFSVVRQCWEKASGRALAAKIIPYHPKDKTAVLREYEALKGLRHPHLA | |
RUO | |
OBSCN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84033 | |
OBSCN (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp666E245/FLJ14124/KIAA1556/KIAA1639/MGC120409/MGC120410/MGC120411/MGC120412/MGC138590/UNC89 | |
OBSCN | |
Recombinant | |
wheat germ expression system |