missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OATL1 Partial ORF (NP_002527.1, 302 a.a. - 410 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004943-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16172155

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 31-10-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a protein with a TBC domain and may function as a Rab GTPase activating protein. This gene was previously known as ornithine aminotransferase-like 1, but has no similarity to ornithine aminotransferase. [provided by RefSeq]

Sequence: YAGPEDGPHLRALHDLLTTYAVTHPQVSYCQGMSDLASPILAVMDHEGHAFVCFCGIMKRLAANFHPDGRAMATKFAHLKLLLRHADPDFYQYLQEAGADDLFFCYRWL
Especificaciones

Especificaciones

NP_002527.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YAGPEDGPHLRALHDLLTTYAVTHPQVSYCQGMSDLASPILAVMDHEGHAFVCFCGIMKRLAANFHPDGRAMATKFAHLKLLLRHADPDFYQYLQEAGADDLFFCYRWL
RUO
TBC1D25
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4943
OATL1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MG81/MGC126866/MGC126868/MGC149731/MGC149732/OATL1
TBC1D25
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales