missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NOPE Partial ORF (NP_066013, 1152 a.a. - 1250 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00057722-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: DLEPEDPLPPEAPDLISGVGDPGQGAAWLDRELGGCELAAPGPDRLTCLPEAASASCSYPDLQPGEVLEETPGDSCQLKSPCPLGASPGLPRSPVSSSAEspecificaciones
NP_066013 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DLEPEDPLPPEAPDLISGVGDPGQGAAWLDRELGGCELAAPGPDRLTCLPEAASASCSYPDLQPGEVLEETPGDSCQLKSPCPLGASPGLPRSPVSSSA | |
RUO | |
IGDCC4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57722 | |
NOPE (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DDM36/FLJ42051/KIAA1628/NOPE | |
IGDCC4 | |
Recombinant | |
wheat germ expression system |