Learn More
ACROBiosystems Human NKp46 / NCR1 / CD335 Protein, Llama IgG2b Fc Tag, low endotoxin
low endotoxin
Marca: ACROBiosystems NC1-H5255-100ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Natural cytotoxicity triggering receptor 1 (NCR1) is also known as Natural killer cell p46-related protein (NK-p46), Lymphocyte antigen 94 homolog (LY94), CD antigen CD335, which belongs to the natural cytotoxicity receptor (NCR) family. NCR1 contains two Ig-like (immunoglobulin-like) domains. NCR1 interacts with CD247 and FCER1G. NCR1 / CD335 may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis.
This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 54.2 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_001138929.2(K82Q) | |
100 μg | |
Gln 22 - Asn 254 | |
NCR1,LY94,CD335,NK-p46,hNKp46 | |
Unconjugated | |
Llama IgG Fc Tag | |
Less than 0.01 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
54.2 kDa | |
Human NKp46, Llama IgG2b Fc Tag, low endotoxin (NC1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession # AAH64806). | |
-20°C | |
NKp46 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |