missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NIPSNAP3B Full-length ORF (NP_060846.2, 1 a.a. - 247 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055335-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
NIPSNAP3B belongs to a family of proteins with putative roles in vesicular trafficking (Buechler et al., 2004 [PubMed 15177564]).[supplied by OMIM]
Sequence: MLVLRSGLTKALASRTLAPQVCSSFATGPRQYDGTFYEFRTYYLKPSNMNAFMENLKKNIHLRTSYSELVGFWSVEFGGRTNKVFHIWKYDNFAHRAEVRKALANCKEWQEQSIIPNLARIDKQETEITYLIPWSKLEKPPKEGVYELAVFQMKPGGPALWGDAFERAINAHVNLGYTKVVGVFHTEYGELNRVHVLWWNESADSRAAGRHKSHEDPRVVAAVRESVNYLVSQQNMLLIPASFSPLKEspecificaciones
NP_060846.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11275/NIPSNAP3 | |
NIPSNAP3B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55335 | |
NIPSNAP3B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLVLRSGLTKALASRTLAPQVCSSFATGPRQYDGTFYEFRTYYLKPSNMNAFMENLKKNIHLRTSYSELVGFWSVEFGGRTNKVFHIWKYDNFAHRAEVRKALANCKEWQEQSIIPNLARIDKQETEITYLIPWSKLEKPPKEGVYELAVFQMKPGGPALWGDAFERAINAHVNLGYTKVVGVFHTEYGELNRVHVLWWNESADSRAAGRHKSHEDPRVVAAVRESVNYLVSQQNMLLIPASFSPLK | |
RUO | |
NIPSNAP3B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |