Learn More
Abnova™ Human NFYC Full-length ORF (NP_055038.2, 1 a.a. - 335 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004802-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes one subunit of a trimeric complex forming a highly conserved transcription factor that binds with high specificity to CCAAT motifs in the promoters of a variety of genes. The encoded protein, subunit C, forms a tight dimer with the B subunit, a prerequisite for subunit A association. The resulting trimer binds to DNA with high specificity and affinity. Subunits B and C each contain a histone-like motif. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MSTEGGFGGTSSSDAQQSLQSFWPRVMEEIRNLTVKDFRVQELPLARIKKIMKLDEDVKMISAEAPVLFAKAAQIFITELTLRAWIHTEDNKRRTLQRNDIAMAITKFDQFDFLIDIVPRDELKPPKRQEEVRQSVTPAEPVQYYFTLAQQPTAVQVQGQQQGQQTTSSTTTIQPGQIIIAQPQQGQTTPVTMQVGEGQQVQIVQAQPQGQAQQAQSGTGQTMQVMQQIITNTGEIQQIPVQLNAGQLQYIRLAQPVSGTQVVQGQIQTLATNAQQITQTEVQQGQQQFSQFTDGQQLYQIQQVTMPAGQDLAQPMFIQSANQPSDGQAPQVTGDEspecificaciones
NP_055038.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CBF-C/CBFC/DKFZp667G242/FLJ45775/H1TF2A/HAP5/HSM/NF-YC | |
NFYC | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4802 | |
NFYC (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSTEGGFGGTSSSDAQQSLQSFWPRVMEEIRNLTVKDFRVQELPLARIKKIMKLDEDVKMISAEAPVLFAKAAQIFITELTLRAWIHTEDNKRRTLQRNDIAMAITKFDQFDFLIDIVPRDELKPPKRQEEVRQSVTPAEPVQYYFTLAQQPTAVQVQGQQQGQQTTSSTTTIQPGQIIIAQPQQGQTTPVTMQVGEGQQVQIVQAQPQGQAQQAQSGTGQTMQVMQQIITNTGEIQQIPVQLNAGQLQYIRLAQPVSGTQVVQGQIQTLATNAQQITQTEVQQGQQQFSQFTDGQQLYQIQQVTMPAGQDLAQPMFIQSANQPSDGQAPQVTGD | |
RUO | |
NFYC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |