missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NFKBIB Full-length ORF (AAH15528, 1 a.a. - 356 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004793-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16168351

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 03-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

NFKB1 (MIM 164011) or NFKB2 (MIM 164012) is bound to REL (MIM 164910), RELA (MIM 164014), or RELB (MIM 604758) to form the NFKB complex. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA, MIM 164008, or NFKBIB), which inactivate NF-kappa-B by trapping it in the cytoplasm. Phosphorylation of serine residues on the I-kappa-B proteins by kinases (IKBKA, MIM 600664 or IKBKB, MIM 603258) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B complex. Activated NFKB complex translocates into the nucleus and binds DNA at kappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or 5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or G purine; and Y is a C or T pyrimidine).[supplied by OMIM]

Sequence: MAGVACLGKAADADEWCDSGLGSLGPDAAAPGGPGLGAELGPGLSWAPLVFGYVTEDGDTALHLAVIHQHEPFLDFLLGFSAGTEYMDLQNDLGQTALHLAAILGETSTVEKLYAAGAGLCVAERRGHTALHLACRVGAHACARALLQPRPRRPREAPDTYLAQGPDRTPDTNHTPVALYPDSDLEKEEEESEEDWKLQLEAENYEGHTPLHVAVIHKDVEMVRLLRDAGADLDKPEPTCGRSPLHLAVEAQAADVLELLLRAGANPAARMYGGRTPLGSAMLRPNPILARLLRAHGAPEPEGEDEKSGPCSSSSDSDSGDEGDEYDDIVVHSSRSQTRLPPTPASKPLPDDPRPV
Especificaciones

Especificaciones

AAH15528
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IKBB/TRIP9
NFKBIB
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4793
NFKBIB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGVACLGKAADADEWCDSGLGSLGPDAAAPGGPGLGAELGPGLSWAPLVFGYVTEDGDTALHLAVIHQHEPFLDFLLGFSAGTEYMDLQNDLGQTALHLAAILGETSTVEKLYAAGAGLCVAERRGHTALHLACRVGAHACARALLQPRPRRPREAPDTYLAQGPDRTPDTNHTPVALYPDSDLEKEEEESEEDWKLQLEAENYEGHTPLHVAVIHKDVEMVRLLRDAGADLDKPEPTCGRSPLHLAVEAQAADVLELLLRAGANPAARMYGGRTPLGSAMLRPNPILARLLRAHGAPEPEGEDEKSGPCSSSSDSDSGDEGDEYDDIVVHSSRSQTRLPPTPASKPLPDDPRPV
RUO
NFKBIB
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales