Learn More
ACROBiosystems Human Neuroligin-4, Y-linked / NLGN4Y Protein, His Tag
Marca: ACROBiosystems NL1-H5228-250UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Neuroligin 4, Y-linked (NLGN4Y) a type I membrane protein that belongs to the family of neuroligins, which are cell adhesion molecules present at the postsynaptic side of the synapse, and may be essential for the formation of functional synapses. Alternatively spliced transcript variants have been found for NLGN4Y gene. NLGN4Y was expressed in fetal and adult brain, prostate, testis, pancreas. NLGN4Y is homologous with its X-linked homolog, NLGN4X.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 73.1 kDa. The protein migrates as 85-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_055708.3 | |
250 µg | |
Gln 44 - Ser 676 | |
Neuroligin-4, Y-linked | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
73.1 kDa | |
Human Neuroligin-4, Y-linked, His Tag (NL1-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 44 - Ser 676 (Accession # Q8NFZ3-1). | |
-20°C | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |