Learn More
ACROBiosystems Human Netrin receptor DCC / DCC Protein, His Tag
MALS verified
Marca: ACROBiosystems NEC-H52H3-1mg
Detalles adicionales : Peso : 0.00010kg
Descripción
Receptor for netrin required for axon guidance. Mediates axon attraction of neuronal growth cones in the developing nervous system upon ligand binding. Its association with UNC5 proteins may trigger signaling for axon repulsion. It also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand. Implicated as a tumor suppressor gene.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 119.6 kDa. The protein migrates as 125-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_005206.2 | |
1 mg | |
Phe 32 - Asn 1097 | |
Netrin receptor DCC,DCC,IGDCC1,netrin receptor DCC | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
119.6 kDa | |
Human Netrin receptor DCC, His Tag (NEC-H52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 32 - Asn 1097 (Accession # P43146-1). | |
-20°C | |
Netrin receptor DCC | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |