Learn More
Abnova™ Human NELL1 Full-length ORF (AAH69674.1, 1 a.a. - 810 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004745-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF)-like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MPMDLILVVWFCVCTARTVVGFGMDPDLQMDIVTELDLVNTTLGVAQVSGMHNASKAFLFQDIEREIHAAPHVSEKLIQLFRNKSEFTILATVQQKPSTSGVILSIRELEHSYFELESSGLRDEIRYHYIHNGKPRTEALPYRMADGQWHKVALSVSASHLLLHVDCNRIYERVIDPPDTNLPPGINLWLGQRNQKHGLFKGIIQDGKIIFMPNGYITQCPNLNHTCPTCSDFLSLVQGIMDLQELLAKMTAKLNYAETRLSQLENCHCEKTCQVSGLLYRDQDSWVDGDHCRNCTCKSGAVECRRMSCPPLNCSPDSLPVHIAGQCCKVCRPKCIYGGKVLAEGQRILTKSCRECRGGVLVKITEMCPPLNCSEKDHILPENQCCRVCRGHNFCAEGPKCGENSECKNWNTKATCECKSGYISVQGDSAYCEDIDECAAKMHYCHANTVCVNLPGLYRCDCVPGYIRVDDFSCTEHDECGSGQHNCDENAICTNTVQGHSCTCKPGYVGNGTICRAFCEEGCRYGGTCVAPNKCVCPSGFTGSHCEKDIDECSEGIIECHNHSRCVNLPGWYHCECRSGFHDDGTYSLSGESCIDIDECALRTHTCWNDSACINLAGGFDCLCPSGPSCSGDCPHEGGLKHNGQVWTLKEDRCSVCSCKDGKIFCRRTACDCQNPSADLFCCPECDTRVTSQCLDQNGHKLYRSGDNWTHSCQQCRCLEGEVDCWPLTCPNLSCEYTAILEGECCPRCVSDPCLADNITYDIRKTCLDSYGVSRLSGSVWTMAGSPCTTCKCKNGRVCCSVDFECLQNNEspecificaciones
AAH69674.1 | |
Liquid | |
4745 | |
NELL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPMDLILVVWFCVCTARTVVGFGMDPDLQMDIVTELDLVNTTLGVAQVSGMHNASKAFLFQDIEREIHAAPHVSEKLIQLFRNKSEFTILATVQQKPSTSGVILSIRELEHSYFELESSGLRDEIRYHYIHNGKPRTEALPYRMADGQWHKVALSVSASHLLLHVDCNRIYERVIDPPDTNLPPGINLWLGQRNQKHGLFKGIIQDGKIIFMPNGYITQCPNLNHTCPTCSDFLSLVQGIMDLQELLAKMTAKLNYAETRLSQLENCHCEKTCQVSGLLYRDQDSWVDGDHCRNCTCKSGAVECRRMSCPPLNCSPDSLPVHIAGQCCKVCRPKCIYGGKVLAEGQRILTKSCRECRGGVLVKITEMCPPLNCSEKDHILPENQCCRVCRGHNFCAEGPKCGENSECKNWNTKATCECKSGYISVQGDSAYCEDIDECAAKMHYCHANTVCVNLPGLYRCDCVPGYIRVDDFSCTEHDECGSGQHNCDENAICTNTVQGHSCTCKPGYVGNGTICRAFCEEGCRYGGTCVAPNKCVCPSGFTGSHCEKDIDECSEGIIECHNHSRCVNLPGWYHCECRSGFHDDGTYSLSGESCIDIDECALRTHTCWNDSACINLAGGFDCLCPSGPSCSGDCPHEGGLKHNGQVWTLKEDRCSVCSCKDGKIFCRRTACDCQNPSADLFCCPECDTRVTSQCLDQNGHKLYRSGDNWTHSCQQCRCLEGEVDCWPLTCPNLSCEYTAILEGECCPRCVSDPCLADNITYDIRKTCLDSYGVSRLSGSVWTMAGSPCTTCKCKNGRVCCSVDFECLQNN | |
RUO | |
NELL1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
116kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ45906/IDH3GL/NRP1 | |
NELL1 | |
Yes | |
wheat germ expression system |