missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NEK9 Partial ORF (AAH09336, 226 a.a. - 326 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00091754-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: AGKGTPLTPPACACSSLQVEVERLQGLVLKCLAEQQKLQQENLQIFTQLQKLNKKLEGGQQVGMHSKGTQTAKEEMEMDPKPDLDSDSWCLLGTDSCRPSLEspecificaciones
AAH09336 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AGKGTPLTPPACACSSLQVEVERLQGLVLKCLAEQQKLQQENLQIFTQLQKLNKKLEGGQQVGMHSKGTQTAKEEMEMDPKPDLDSDSWCLLGTDSCRPSL | |
RUO | |
NEK9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
91754 | |
NEK9 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434D0935/MGC138306/MGC16714/NERCC/NERCC1/Nek8 | |
NEK9 | |
Recombinant | |
wheat germ expression system |