missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NEDD4L Partial ORF (AAH32597, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023327-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MATGLGEPVYGLSEDEGESRILRVKVVSGIDLAKKDIFGASDPYVKLSLYVADENRELALVQTKTIKKTLNPKWNEEFYFRVNPSNHRLLFEVFDENRLTEspecificaciones
AAH32597 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATGLGEPVYGLSEDEGESRILRVKVVSGIDLAKKDIFGASDPYVKLSLYVADENRELALVQTKTIKKTLNPKWNEEFYFRVNPSNHRLLFEVFDENRLT | |
RUO | |
NEDD4L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23327 | |
NEDD4L (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ33870/KIAA0439/NEDD4-2/RSP5/hNedd4-2 | |
NEDD4L | |
Recombinant | |
wheat germ expression system |