Learn More
Abnova™ Human NECAP2 Full-length ORF (NP_060560.1, 1 a.a. - 263 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055707-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene likely encodes a member of the adaptin-ear-binding coat-associated protein family. Studies of a similar protein in rat suggest a role in clathrin-mediated endocytosis. Alternatively spliced transcript variants have been described. [provided by RefSeq]
Sequence: MEESGYESVLCVKPDVHVYRIPPRATNRGYRAAEWQLDQPSWSGRLRITAKGQMAYIKLEDRTSGELFAQAPVDQFPGTAVESVTDSSRYFVIRIEDGNGRRAFIGIGFGDRGDAFDFNVALQDHFKWVKQQCEFAKQAQNPDQGPKLDLGFKEGQTIKLNIANMKKKEGAAGNPRVRPASTGGLSLLPPPPGGKTSTLIPPPGEQLAVGGSLVQPAVAPSSGGAPVPWPQPNPATADIWGDFTKSTGSTSSQTQPGTGWVQFEspecificaciones
NP_060560.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
54.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10420/RP4-798A10.1 | |
NECAP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55707 | |
NECAP2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEESGYESVLCVKPDVHVYRIPPRATNRGYRAAEWQLDQPSWSGRLRITAKGQMAYIKLEDRTSGELFAQAPVDQFPGTAVESVTDSSRYFVIRIEDGNGRRAFIGIGFGDRGDAFDFNVALQDHFKWVKQQCEFAKQAQNPDQGPKLDLGFKEGQTIKLNIANMKKKEGAAGNPRVRPASTGGLSLLPPPPGGKTSTLIPPPGEQLAVGGSLVQPAVAPSSGGAPVPWPQPNPATADIWGDFTKSTGSTSSQTQPGTGWVQF | |
RUO | |
NECAP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |