Learn More
Abnova™ Human NDUFB3 Partial ORF (NP_002482, 13 a.a. - 65 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00004709-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16171735
  • 349.00€ / 10 microgramos
Fecha estimada de envío: 07-11-2023
para ver el stock
Descripción
Descripción
The multisubunit NADH:ubiquinone oxidoreductase (complex I) is the first enzyme complex in the electron transport chain of mitochondria. See NDUFA2 (MIM 602137).[supplied by OMIM]
Sequence: KMELPDYRQWKIEGTPLETIQKKLAAKGLRDPWGRNEAWRYMGGFAKSVSFSD
Especificaciones
Especificaciones
NP_002482
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.57kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KMELPDYRQWKIEGTPLETIQKKLAAKGLRDPWGRNEAWRYMGGFAKSVSFSD
RUO
NDUFB3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4709
NDUFB3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
B12
NDUFB3
Recombinant
wheat germ expression system