Learn More
Abnova™ Human NDST2 Partial ORF (NP_003626.1, 172 a.a. - 270 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008509-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the N-deacetylase/N-sulfotransferase subfamily of the sulfotransferase 1 proteins. The encoded enzyme has dual functions in processing glucosamine and heparin polymers, including N-deacetylation and N-sulfation. The encoded protein may be localized to the Golgi. [provided by RefSeq]
Sequence: FRAHEHSLLSAQLKGFPLFLHSNLGLRDYQVNPSAPLLHLTRPSRLEPGPLPGDDWTIFQSNHSTYEPVLLASLRPAEPAVPGPVLRRARLPTVVQDLGEspecificaciones
NP_003626.1 | |
Liquid | |
8509 | |
NDST2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSST2/MGC129697/MGC129705/NCST2/NST2 | |
NDST2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FRAHEHSLLSAQLKGFPLFLHSNLGLRDYQVNPSAPLLHLTRPSRLEPGPLPGDDWTIFQSNHSTYEPVLLASLRPAEPAVPGPVLRRARLPTVVQDLG | |
RUO | |
NDST2 | |
Wheat Germ (in vitro) | |
GST |