missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NCOA6 Partial ORF (NP_054790, 1954 a.a. - 2063 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00023054-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16166892

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 03-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a transcriptional coactivator that can interact with nuclear hormone receptors to enhance their transcriptional activator functions. The encoded protein has been shown to be involved in the hormone-dependent coactivation of several receptors, including prostanoid, retinoid, vitamin D3, thyroid hormone, and steroid receptors. The encoded protein may also act as a general coactivator since it has been shown to interact with some basal transcription factors, histone acetyltransferases, and methyltransferases. [provided by RefSeq]

Sequence: VGSHPELLPSIAPSQNLVSKETSTTALQASVARPELEVNAAIVSGQSSEPKEIVEKSKIPGRRNSRTEEPTVASESVENGHRKRSSRPASASSSTKDITSAVQSKRRKSK
Especificaciones

Especificaciones

NP_054790
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VGSHPELLPSIAPSQNLVSKETSTTALQASVARPELEVNAAIVSGQSSEPKEIVEKSKIPGRRNSRTEEPTVASESVENGHRKRSSRPASASSSTKDITSAVQSKRRKSK
RUO
NCOA6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
23054
NCOA6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AIB3/ANTP/ASC2/HOX1.1/HOXA7/KIAA0181/NRC/PRIP/RAP250/TRBP
NCOA6
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales