Learn More
Abnova™ Human NCK2 Full-length ORF (NP_001004720.1, 1 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008440-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the NCK family of adaptor proteins. The protein contains three SH3 domains and one SH2 domain. The protein has no known catalytic function but has been shown to bind and recruit various proteins involved in the regulation of receptor protein tyrosine kinases. It is through these regulatory activities that this protein is believed to be involved in cytoskeletal reorganization. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MTEEVIVIAKWDYTAQQDQELDIKKNERLWLLDDSKTWWRVRNAANRTGYVPSNYVERKNSLKKGSLVKNLKDTLGLGKTRRKTSARDASPTPSTDAEYPANGSGADRIYDLNIPAFVKFAYVAEREDELSLVKGSRVTVMEKCSDGWWRGSYNGQIGWFPSNYVLEEVDEAAAESPSFLSLRKGASLSNGQGSRVLHVVQTLYPFSSVTEEELNFEKGETMEVIEKPENDPEWWKCKNARGQVGLVPKNYVVVLSDGPALHPAHAPQISYTGPSSSGRFAGREWYYGNVTRHQAECALNERGVEGDFLIRDSESSPSDFSVSLKASGKNKHFKVQLVDNVYCIGQRRFHTMDELVEHYKKAPIFTSEHGEKLYLVRALQEspecificaciones
NP_001004720.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
69.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GRB4/NCKbeta | |
NCK2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8440 | |
NCK2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTEEVIVIAKWDYTAQQDQELDIKKNERLWLLDDSKTWWRVRNAANRTGYVPSNYVERKNSLKKGSLVKNLKDTLGLGKTRRKTSARDASPTPSTDAEYPANGSGADRIYDLNIPAFVKFAYVAEREDELSLVKGSRVTVMEKCSDGWWRGSYNGQIGWFPSNYVLEEVDEAAAESPSFLSLRKGASLSNGQGSRVLHVVQTLYPFSSVTEEELNFEKGETMEVIEKPENDPEWWKCKNARGQVGLVPKNYVVVLSDGPALHPAHAPQISYTGPSSSGRFAGREWYYGNVTRHQAECALNERGVEGDFLIRDSESSPSDFSVSLKASGKNKHFKVQLVDNVYCIGQRRFHTMDELVEHYKKAPIFTSEHGEKLYLVRALQ | |
RUO | |
NCK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |