Learn More
Abnova™ Human NCAPH Full-length ORF (AAH24211.1, 1 a.a. - 741 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023397-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization. [provided by RefSeq]
Sequence: MGPPGPALPATMNNSSSETRGHPHSASSPSERVFPMPLPRKAPLNIPGTPVLEDFPQNDDEKERLQRRRSRVFDLQFSTDSPRLLASPSSRSIDISATIPKFTNTQITEHYSTCIKLSTENKITTKNAFGLHLIDFMSEILKQKDTEPTNFKVAAGTLDASTKIYAVRVDAVHADVYRVLGGLGKDAPSLEEVEGHVADGSATEMGTTKKAVKPKKKHLHRTIEQNINNLNVSEADRKCEIDPMFQKTAASFDECSTAGVFLSTLHCQDYRSELLFPSDVQTLSTGEPLELPELGCVEMTDLKAPLQQCAEDRQICPSLAGFQFTQWDSETHNESVSALVDKFKKNDQVFDINAEVDESDCGDFPDGSLGDDFDANDEPDHTAVGDHEEFRSWKEPCQVQSCQEEMISLGDGDIRTMCPLLSMKPGEYSYFSPRTMSMWAGPDHWRFRPRRKQDAPSQSENKKKSTKKDFEIDFEDDIDFDVYFRKTKAATILTKSTLENQNWRATTLPTDFNYNVDTLVQLHLKPGTRLLKMAQGHRAETEHYEEIEDYDYNNPNDTSNFCPGLQAADSDDEDLDDLFVGPVGNSDLSPYPCHPPKTAQQNGDTPEAQGLDITTYGESNLVAEPQKVNKIEIHYAKTAKKMDMKKLKQSMWSLLTALSGKEADAEANHREAGKEAALAEVADEKMLSGLTKDLQRSLPPVMAQNLSIPLAFACLLHLANEKNLKLEGTEDLSDVLVRQGDEspecificaciones
AAH24211.1 | |
Liquid | |
23397 | |
NCAPH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGPPGPALPATMNNSSSETRGHPHSASSPSERVFPMPLPRKAPLNIPGTPVLEDFPQNDDEKERLQRRRSRVFDLQFSTDSPRLLASPSSRSIDISATIPKFTNTQITEHYSTCIKLSTENKITTKNAFGLHLIDFMSEILKQKDTEPTNFKVAAGTLDASTKIYAVRVDAVHADVYRVLGGLGKDAPSLEEVEGHVADGSATEMGTTKKAVKPKKKHLHRTIEQNINNLNVSEADRKCEIDPMFQKTAASFDECSTAGVFLSTLHCQDYRSELLFPSDVQTLSTGEPLELPELGCVEMTDLKAPLQQCAEDRQICPSLAGFQFTQWDSETHNESVSALVDKFKKNDQVFDINAEVDESDCGDFPDGSLGDDFDANDEPDHTAVGDHEEFRSWKEPCQVQSCQEEMISLGDGDIRTMCPLLSMKPGEYSYFSPRTMSMWAGPDHWRFRPRRKQDAPSQSENKKKSTKKDFEIDFEDDIDFDVYFRKTKAATILTKSTLENQNWRATTLPTDFNYNVDTLVQLHLKPGTRLLKMAQGHRAETEHYEEIEDYDYNNPNDTSNFCPGLQAADSDDEDLDDLFVGPVGNSDLSPYPCHPPKTAQQNGDTPEAQGLDITTYGESNLVAEPQKVNKIEIHYAKTAKKMDMKKLKQSMWSLLTALSGKEADAEANHREAGKEAALAEVADEKMLSGLTKDLQRSLPPVMAQNLSIPLAFACLLHLANEKNLKLEGTEDLSDVLVRQGD | |
RUO | |
NCAPH | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
108.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRRN1/CAP-H/HCAP-H | |
NCAPH | |
Yes | |
wheat germ expression system |