Learn More
Abnova™ Human NAV1 Full-length ORF (AAH07523.1, 1 a.a. - 559 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00089796-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene belongs to the neuron navigator family and is expressed predominantly in the nervous system. The encoded protein contains coiled-coil domains and a conserved AAA domain characteristic for ATPases associated with a variety of cellular activities. This gene is similar to unc-53, a Caenorhabditis elegans gene involved in axon guidance. The exact function of this gene is not known. [provided by RefSeq]
Sequence: MKLTDIRLEALNSAHQLDQLRETMHNMQLEVDLLKAENDRLKVAPGPSSGSTPGQVPGSSALSSPRRSLGLALTHSFGPSLADTDLSPMDGISTCGPKEEVTLRVVVRMPPQHIIKGDLKQQEFFLGCSKVSGKVDWKMLDEAVFQVFKDYISKMDPASTLGLSTESIHGYSISHVKRVLDAEPPEMPPCRRGVNNISVSLKGLKEKCVDSLVFETLIPKPMMQHYISLLLKHRRLVLSGPSGTGKTYLTNRLAEYLVERSGREVTEGIVSTFNMHQQSCKDLQLYLSNLANQIDRETGIGDVPLVILLDDLSEAGSISELVNGALTCKYHKCPYIIGTTNQPVKMTPNHGLHLSFRMLTFSNNVEPANGFLVRYLRRKLVESDSDINANKEELLRVLDWVPKLWYHLHTFLEKHSTSDFLIGPCFFLSCPIGIEDFRTWFIDLWNNSIIPYLQEGAKDGIKVHGQKAAWEDPVEWVRDTLPWPSAQQDQSKLYHLPPPTVGPHSIASPPEDRTVKDSTPSSLDSDPLMAMLLKLQEAANYIESPDRETILDPNLQATLEspecificaciones
AAH07523.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
88.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781D0314/FLJ12560/FLJ14203/KIAA1151/MGC14961/POMFIL3/steerin-1 | |
NAV1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
89796 | |
NAV1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKLTDIRLEALNSAHQLDQLRETMHNMQLEVDLLKAENDRLKVAPGPSSGSTPGQVPGSSALSSPRRSLGLALTHSFGPSLADTDLSPMDGISTCGPKEEVTLRVVVRMPPQHIIKGDLKQQEFFLGCSKVSGKVDWKMLDEAVFQVFKDYISKMDPASTLGLSTESIHGYSISHVKRVLDAEPPEMPPCRRGVNNISVSLKGLKEKCVDSLVFETLIPKPMMQHYISLLLKHRRLVLSGPSGTGKTYLTNRLAEYLVERSGREVTEGIVSTFNMHQQSCKDLQLYLSNLANQIDRETGIGDVPLVILLDDLSEAGSISELVNGALTCKYHKCPYIIGTTNQPVKMTPNHGLHLSFRMLTFSNNVEPANGFLVRYLRRKLVESDSDINANKEELLRVLDWVPKLWYHLHTFLEKHSTSDFLIGPCFFLSCPIGIEDFRTWFIDLWNNSIIPYLQEGAKDGIKVHGQKAAWEDPVEWVRDTLPWPSAQQDQSKLYHLPPPTVGPHSIASPPEDRTVKDSTPSSLDSDPLMAMLLKLQEAANYIESPDRETILDPNLQATL | |
RUO | |
NAV1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |