Learn More
Abnova™ Human NAP1L2 Full-length ORF (AAH26325.1, 1 a.a. - 460 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004674-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the nucleosome assembly protein (NAP) family. The function of this family member is unknown; however, mouse studies suggest that it represents a class of tissue-specific factors interacting with chromatin to regulate neuronal cell proliferation. [provided by RefSeq]
Sequence: MAKSENRKELSESSQEEAGNQIMVEGLGEHLERGEDAAAGLGDDGKCGEEAAAGLGEEGENGEDTAAGSGEDGKKGGDTDEDSEADRPKGLIGYVLDTDFVESLPVKVKYRVLALKKLQTRAANLESKFLREFHDIERKFAEMYQPLLEKRRQIINAIYEPTEEECEYKSDSEDCDDEEMCHEEMYGNEEGMVHEYVDEDDGYEDYYYDYAVEEEEEEEEEDDIEATGEENKEEEDPKGIPDFWLTVLKNVDTLTPLIKKYDEPILKLLTDIKVKLSDPGEPLSFTLEFHFKPNEYFKNELLTKTYVLKSKLAYYDPHPYRGTAIEYSTGCEIDWNEGKNVTLKTIKKKQKHRIWGTIRTVTEDFPKDSFFNFFSPHGITSNGRDGNDDFLLGHNLRTYIIPRSVLFFSGDALESQQEGVVREVNDAIYDKIIYDNWMAAIEEVKACCKNLEALVEDIDREspecificaciones
AAH26325.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BPX/MGC26243 | |
NAP1L2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4674 | |
NAP1L2 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAKSENRKELSESSQEEAGNQIMVEGLGEHLERGEDAAAGLGDDGKCGEEAAAGLGEEGENGEDTAAGSGEDGKKGGDTDEDSEADRPKGLIGYVLDTDFVESLPVKVKYRVLALKKLQTRAANLESKFLREFHDIERKFAEMYQPLLEKRRQIINAIYEPTEEECEYKSDSEDCDDEEMCHEEMYGNEEGMVHEYVDEDDGYEDYYYDYAVEEEEEEEEEDDIEATGEENKEEEDPKGIPDFWLTVLKNVDTLTPLIKKYDEPILKLLTDIKVKLSDPGEPLSFTLEFHFKPNEYFKNELLTKTYVLKSKLAYYDPHPYRGTAIEYSTGCEIDWNEGKNVTLKTIKKKQKHRIWGTIRTVTEDFPKDSFFNFFSPHGITSNGRDGNDDFLLGHNLRTYIIPRSVLFFSGDALESQQEGVVREVNDAIYDKIIYDNWMAAIEEVKACCKNLEALVEDIDR | |
RUO | |
NAP1L2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |