Learn More
Abnova™ Human NADSYN1 Partial ORF (NP_004981, 609 a.a. - 706 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055191-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Nicotinamide adenine dinucleotide (NAD) is a coenzyme in metabolic redox reactions, a precursor for several cell signaling molecules, and a substrate for protein posttranslational modifications. NAD synthetase (EC 6.3.5.1) catalyzes the final step in the biosynthesis of NAD from nicotinic acid adenine dinucleotide (NaAD).[supplied by OMIM]
Sequence: KMGPYSMFCKLLGMWRHICTPRQVADKVKRFFSKYSMNRHKMTTLTPAYHAENYSPEDNRFDLRPFLYNTSWPWQFRCIENQVLQLERAEPQSLDGVDEspecificaciones
NP_004981 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KMGPYSMFCKLLGMWRHICTPRQVADKVKRFFSKYSMNRHKMTTLTPAYHAENYSPEDNRFDLRPFLYNTSWPWQFRCIENQVLQLERAEPQSLDGVD | |
RUO | |
NADSYN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55191 | |
NADSYN1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10631/FLJ36703/FLJ40627 | |
NADSYN1 | |
Recombinant | |
wheat germ expression system |