missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MYO9B Partial ORF (AAH18108, 201 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004650-Q01.25ug
Weitere Details : Peso : 0.00010kg
Beschreibung
Sequence: CPDNSDPLTSMKDVLKITTCVEMLIKEQMRKYKVKMEEISQLEAAESIAFRRLSLLRQNAPWPLKLGFSSPYEGVLNKSPKTRDIQEEELSpezifikation
AAH18108 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CPDNSDPLTSMKDVLKITTCVEMLIKEQMRKYKVKMEEISQLEAAESIAFRRLSLLRQNAPWPLKLGFSSPYEGVLNKSPKTRDIQEEEL | |
RUO | |
MYO9B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4650 | |
MYO9B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CELIAC4/MYR5 | |
MYO9B | |
Recombinant | |
wheat germ expression system |