missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MYB Partial ORF (AAH64955, 541 a.a. - 640 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004602-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16108051

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 12-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a transcription factor that is a member of the MYB family of transcription factor genes. The protein contains three domains, an N-terminal DNA-binding domain, a central transcriptional activation domain and a C-terminal domain involved in transcriptional repression. This protein plays an essential role in the regulation of hematopoiesis and may play a role in tumorigenesis. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: FCSHHWEGDSLNTQLFTQTSPVADAPNILTSSVLMAPASEDEDNVLKAFTVPKNRSLASPLQPCSSTWEPASCGKMEEQMTSSSQARKYVNAFSARTLVM
Especificaciones

Especificaciones

AAH64955
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FCSHHWEGDSLNTQLFTQTSPVADAPNILTSSVLMAPASEDEDNVLKAFTVPKNRSLASPLQPCSSTWEPASCGKMEEQMTSSSQARKYVNAFSARTLVM
RUO
MYB
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4602
MYB (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Cmyb/c-myb/c-myb_CDS/efg
MYB
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales