Learn More
Abnova™ Human MUSK Partial ORF (NP_005583, 301 a.a. - 400 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004593-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Intercellular communication is often mediated by receptors on the surface of one cell that recognize and are activated by specific protein ligands released by other cells. Members of one class of cell surface receptors, receptor tyrosine kinases (RTKs), are characterized by having a cytoplasmic domain containing intrinsic tyrosine kinase activity. This kinase activity is regulated by the binding of a cognate ligand to the extracellular portion of the receptor. DeChiara et al. (1996) [PubMed 8653786] noted that the RTKs, known to be expressed in cell type-specific fashions, play a role critical for the growth and differentiation of those cell types. For example, members of the neural-specific TRK family that recognize nerve growth factor are absolutely required for the survival and development of discrete neuronal subpopulations, and the receptor tyrosine kinases TIE1 (MIM 600222) and TIE2 (MIM 600221) play a critical role in the development of normal blood vessels.[supplied by OMIM]
Sequence: ISIAEWSKPQKDNKGYCAQYRGEVCNAVLAKDALVFLNTSYADPEEAQELLVHTAWNELKVVSPVCRPAAEALLCNHIFQECSPGVVPTPIPICREYCLAEspecificaciones
NP_005583 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ISIAEWSKPQKDNKGYCAQYRGEVCNAVLAKDALVFLNTSYADPEEAQELLVHTAWNELKVVSPVCRPAAEALLCNHIFQECSPGVVPTPIPICREYCLA | |
RUO | |
MUSK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4593 | |
MUSK (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126323/MGC126324 | |
MUSK | |
Recombinant | |
wheat germ expression system |