Learn More
Abnova™ Human MSX1 Partial ORF (NP_002439.1, 216 a.a. - 297 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004487-Q01.10ug
Detalles adicionales : Peso : 0.02000kg
Descripción
This gene encodes a member of the muscle segment homeobox gene family. The encoded protein functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. It may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition. Mutations in this gene, which was once known as homeobox 7, have been associated with nonsyndromic cleft lip with or without cleft palate 5, Witkop syndrome, Wolf-Hirschom syndrome, and autosomoal dominant hypodontia. [provided by RefSeq]
Sequence: NRRAKAKRLQEAELEKLKMAAKPMLPPAAFGLSFPLGGPAAVAAAAGASLYGASGPFQRAALPVAPVGLYTAHVGYSMYHLTEspecificaciones
NP_002439.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.65kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
NRRAKAKRLQEAELEKLKMAAKPMLPPAAFGLSFPLGGPAAVAAAAGASLYGASGPFQRAALPVAPVGLYTAHVGYSMYHLT | |
RUO | |
MSX1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4487 | |
MSX1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX7/HYD1 | |
MSX1 | |
Recombinant | |
wheat germ expression system |