Learn More
Abnova™ Human MRPL20 Full-length ORF (NP_060441.2, 1 a.a. - 149 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055052-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found on chromosome 21q. [provided by RefSeq]
Sequence: MVFLTAQLWLRNRVTDRYFRIQEVLKHARHFRGRKNRCYRLAVRTVIRAFVKCTKARYLKKKNMRTLWINRITAASQEHGLKYPALIGNLVKCQVELNRKVLADLAIYEPKTFKSLAALASRRRHEGFAAALGDGKEPEGIFSRVVQYHEspecificaciones
NP_060441.2 | |
Liquid | |
55052 | |
MRPL20 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVFLTAQLWLRNRVTDRYFRIQEVLKHARHFRGRKNRCYRLAVRTVIRAFVKCTKARYLKKKNMRTLWINRITAASQEHGLKYPALIGNLVKCQVELNRKVLADLAIYEPKTFKSLAALASRRRHEGFAAALGDGKEPEGIFSRVVQYH | |
RUO | |
MRPL20 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
43.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
L20mt/MGC4779/MGC74465/MRP-L20 | |
MRPL20 | |
Yes | |
wheat germ expression system |