Learn More
Abnova™ Human MOS Full-length ORF (NP_005363.1, 1 a.a. - 346 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004342-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
MOS is a serine/threonine kinase that activates the MAP kinase cascade through direct phosphorylation of the MAP kinase activator MEK (MAP2K1; MIM 176872) (Prasad et al., 2008 [PubMed 18246541]).[supplied by OMIM]
Sequence: MPSPLALRPYLRSEFSPSVDARPCSSPSELPAKLLLGATLPRAPRLPRRLAWCSIDWEQVCLLQRLGAGGFGSVYKATYRGVPVAIKQVNKCTKNRLASRRSFWAELNVARLRHDNIVRVVAASTRTPAGSNSLGTIIMEFGGNVTLHQVIYGAAGHPEGDAGEPHCRTGGQLSLGKCLKYSLDVVNGLLFLHSQSIVHLDLKPANILISEQDVCKISDFGCSEKLEDLLCFQTPSYPLGGTYTHRAPELLKGEGVTPKADIYSFAITLWQMTTKQAPYSGERQHILYAVVAYDLRPSLSAAVFEDSLPGQRLGDVIQRCWRPSAAQRPSARLLLVDLTSLKAELGEspecificaciones
NP_005363.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119962/MGC119963/MSV | |
MOS | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4342 | |
MOS (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPSPLALRPYLRSEFSPSVDARPCSSPSELPAKLLLGATLPRAPRLPRRLAWCSIDWEQVCLLQRLGAGGFGSVYKATYRGVPVAIKQVNKCTKNRLASRRSFWAELNVARLRHDNIVRVVAASTRTPAGSNSLGTIIMEFGGNVTLHQVIYGAAGHPEGDAGEPHCRTGGQLSLGKCLKYSLDVVNGLLFLHSQSIVHLDLKPANILISEQDVCKISDFGCSEKLEDLLCFQTPSYPLGGTYTHRAPELLKGEGVTPKADIYSFAITLWQMTTKQAPYSGERQHILYAVVAYDLRPSLSAAVFEDSLPGQRLGDVIQRCWRPSAAQRPSARLLLVDLTSLKAELG | |
RUO | |
MOS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |