missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MNT Partial ORF (NP_064706, 473 a.a. - 582 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004335-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16167891

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 11-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The Myc/Max/Mad network comprises a group of transcription factors that co-interact to regulate gene-specific transcriptional activation or repression. This gene encodes a protein member of the Myc/Max/Mad network. This protein has a basic-Helix-Loop-Helix-zipper domain (bHLHzip) with which it binds the canonical DNA sequence CANNTG, known as the E box, following heterodimerization with Max proteins. This protein is likely a transcriptional repressor and an antagonist of Myc-dependent transcriptional activation and cell growth. This protein represses transcription by binding to DNA binding proteins at its N-terminal Sin3-interaction domain. [provided by RefSeq]

Sequence: SAPSPAVQLAPATPPIGHITVHPATLNHVAHLGSQLPLYPQPVAVSHIAHTLSHQQVNGTAGLGPPATVMAKPAVGAQVVHHPQLVGQTVLNPVTMVTMPSFPVSTLKLA
Especificaciones

Especificaciones

NP_064706
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SAPSPAVQLAPATPPIGHITVHPATLNHVAHLGSQLPLYPQPVAVSHIAHTLSHQQVNGTAGLGPPATVMAKPAVGAQVVHHPQLVGQTVLNPVTMVTMPSFPVSTLKLA
RUO
MNT
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4335
MNT (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MAD6/MXD6/ROX/bHLHd3
MNT
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales