Learn More
Abnova™ Human MN1 Partial ORF (NP_002421, 1212 a.a. - 1320 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004330-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Meningioma 1 (MN1) contains two sets of CAG repeats. It is disrupted by a balanced translocation (4;22) in a meningioma, and its inactivation may contribute to meningioma 32 pathogenesis. [provided by RefSeq]
Sequence: STIDLDSLMAEHSAAWYMPADKALVDSADDDKTLAPWEKAKPQNPNSKEAHDLPANKASASQPGSHLQCLSVHCTDDVGDAKARASVPTWRSLHSDISNRFGTFVAALTEspecificaciones
NP_002421 | |
Liquid | |
4330 | |
MN1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGCR/MGCR1/MGCR1-PEN/dJ353E16.2 | |
MN1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
STIDLDSLMAEHSAAWYMPADKALVDSADDDKTLAPWEKAKPQNPNSKEAHDLPANKASASQPGSHLQCLSVHCTDDVGDAKARASVPTWRSLHSDISNRFGTFVAALT | |
RUO | |
MN1 | |
Wheat Germ (in vitro) | |
GST |