missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MMAB Partial ORF (NP_443077.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00326625-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16172787

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 07-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a protein that catalyzes the final step in the conversion of vitamin B(12) into adenosylcobalamin (AdoCbl), a vitamin B12-containing coenzyme for methylmalonyl-CoA mutase. Mutations in the gene are the cause of vitamin B12-dependent methylmalonic aciduria linked to the cblB complementation group. [provided by RefSeq]

Sequence: MAVCGLGSRLGLGSRLGLRGCFGAARLLYPRFQSRGPQGVEDGDRPQPSSKTPRIPKIYTKTGDKGFSSTFTGERRPKDDQVFEAVGTTDELSSAIGFAL
Especificaciones

Especificaciones

NP_443077.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MAVCGLGSRLGLGSRLGLRGCFGAARLLYPRFQSRGPQGVEDGDRPQPSSKTPRIPKIYTKTGDKGFSSTFTGERRPKDDQVFEAVGTTDELSSAIGFAL
RUO
MMAB
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
326625
MMAB (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATR/MGC20496
MMAB
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales