missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MIB2 Partial ORF (AAH37542, 381 a.a. - 480 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00142678-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: IGDLDTVKRLQAGHGEWTDDMAPALGRVGKVVKVFGDGNLRVAVAGQRWTFSPSCLVAYRPEEDANLDVAERARENKSSLSVALDKLRAQKSDPEHPGRLEspecificaciones
AAH37542 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IGDLDTVKRLQAGHGEWTDDMAPALGRVGKVVKVFGDGNLRVAVAGQRWTFSPSCLVAYRPEEDANLDVAERARENKSSLSVALDKLRAQKSDPEHPGRL | |
RUO | |
MIB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
142678 | |
MIB2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20648/FLJ39787/ZZANK1/ZZZ5 | |
MIB2 | |
Recombinant | |
wheat germ expression system |