Learn More
Abnova™ Human MHC2TA Partial ORF (NP_000237, 39 a.a. - 148 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004261-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein with an acidic transcriptional activation domain, 4 LRRs (leucine-rich repeats) and a GTP binding domain. The protein is located in the nucleus and acts as a positive regulator of class II major histocompatibility complex gene transcription, and is referred to as the “master control factor” for the expression of these genes. The protein also binds GTP and uses GTP binding to facilitate its own transport into the nucleus. Once in the nucleus it does not bind DNA but rather uses an intrinsic acetyltransferase (AT) activity to act in a coactivator-like fashion. Mutations in this gene have been associated with bare lymphocyte syndrome type II (also known as hereditary MHC class II deficiency or HLA class II-deficient combined immunodeficiency), increased susceptibility to rheumatoid arthritis, multiple sclerosis, and possibly myocardial infarction. [provided by RefSeq]
Sequence: NSDADPLCLYHFYDQMDLAGEEEIELYSEPDTDTINCDQFSRLLCDMEGDEETREAYANIAELDQYVFQDSQLEGLSKDIFKHIGPDEVIGESMEMPAEVGQKSQKRPFPEspecificaciones
NP_000237 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NSDADPLCLYHFYDQMDLAGEEEIELYSEPDTDTINCDQFSRLLCDMEGDEETREAYANIAELDQYVFQDSQLEGLSKDIFKHIGPDEVIGESMEMPAEVGQKSQKRPFP | |
RUO | |
CIITA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4261 | |
MHC2TA (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C2TA/CIITAIV/MHC2TA/NLRA | |
CIITA | |
Recombinant | |
wheat germ expression system |