missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MEN1 Partial ORF (NP_000235, 506 a.a. - 615 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004221-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16167751

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 03-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes menin, a putative tumor suppressor associated with a syndrome known as multiple endocrine neoplasia type 1. In vitro studies have shown menin is localized to the nucleus, possesses two functional nuclear localization signals, and inhibits transcriptional activation by JunD, however, the function of this protein is not known. Two messages have been detected on northern blots but the larger message has not been characterized. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: DKGLGTGQGAVSGPPRKPPGTVAGTARGPEGGSTAQVPAPAASPPPEGPVLTFQSEKMKGMKELLVATKINSSAIKLQLTAQSQVQMKKQKVSTPSDYTLSFLKRQRKGL
Especificaciones

Especificaciones

NP_000235
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DKGLGTGQGAVSGPPRKPPGTVAGTARGPEGGSTAQVPAPAASPPPEGPVLTFQSEKMKGMKELLVATKINSSAIKLQLTAQSQVQMKKQKVSTPSDYTLSFLKRQRKGL
RUO
MEN1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4221
MEN1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MEAI/SCG2
MEN1
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales