Learn More
Abnova™ Human MEFV Partial ORF (NP_000234, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004210-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein, also known as pyrin or marenostrin, that is an important modulator of innate immunity. Mutations in this gene are associated with Mediterranean fever, a hereditary periodic fever syndrome. [provided by RefSeq]
Sequence: MAKTPSDHLLSTLEELVPYDFEKFKFKLQNTSVQKEHSRIPRSQIQRARPVKMATLLVTYYGEEYAVQLTLQVLRAINQRLLAEELHRAAIQEYSTQENGTDDSAASSSLEspecificaciones
NP_000234 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAKTPSDHLLSTLEELVPYDFEKFKFKLQNTSVQKEHSRIPRSQIQRARPVKMATLLVTYYGEEYAVQLTLQVLRAINQRLLAEELHRAAIQEYSTQENGTDDSAASSSL | |
RUO | |
MEFV | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4210 | |
MEFV (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FMF/MEF/MGC126560/MGC126586/TRIM20 | |
MEFV | |
Recombinant | |
wheat germ expression system |