missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MED4 Full-length ORF (AAH05189, 1 a.a. - 270 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00029079-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MAASSSGEKEKERLGGGLGVAGGNSTRERLLSALEDLEVLSRELIEMLAISRNQKLLQAGEENQVLELLIHRDGEFQELMKLALNQGKIHHEMQVLEKEVEKRDGDIQQLQKQLKEAEQILATAVYQAKEKLKSIEKARKGAISSEEIIKYAHRISASNAVCAPLTWVPGDPRRPYPTDLEMRSGLLGQMNNPSTNGVNGHLPGDALAAGRLPDVLAPQYPWQSNDMSMNMLPPNHSSDFLLEPPGHNKEDEDDVEIMSTDSSSSSSESDEspecificaciones
AAH05189 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.44kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DRIP36/FLJ10956/HSPC126/RP11-90M2.2/TRAP36/VDRIP | |
MED4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
29079 | |
MED4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAASSSGEKEKERLGGGLGVAGGNSTRERLLSALEDLEVLSRELIEMLAISRNQKLLQAGEENQVLELLIHRDGEFQELMKLALNQGKIHHEMQVLEKEVEKRDGDIQQLQKQLKEAEQILATAVYQAKEKLKSIEKARKGAISSEEIIKYAHRISASNAVCAPLTWVPGDPRRPYPTDLEMRSGLLGQMNNPSTNGVNGHLPGDALAAGRLPDVLAPQYPWQSNDMSMNMLPPNHSSDFLLEPPGHNKEDEDDVEIMSTDSSSSSSESD | |
RUO | |
MED4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |