Learn More
Abnova™ Human MAP3K2 Partial ORF (NP_006600, 111 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
341.70€ - 518.16€
Especificaciones
Nombre MAP3K2 (Human) Recombinant Protein (Q01)
Recombinante Yes
Para utilizar con (aplicación) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Peso molecular 35.53kDa
Requisitos de almacenamiento Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Ver más especificaciones
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16115652
Ver documentos Detalles de la promoción
Abnova™
H00010746-Q01.25UG
25 ug
518.16€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Envío estimado: 08-03-2022
16105652
Ver documentos Detalles de la promoción
Abnova™
H00010746-Q01.10UG
10 ug
341.70€
10 microgramos
Ver productos alternativos
Ahorre hasta 
»
Envío estimado: 08-03-2022
Descripción
Descripción
The protein encoded by this gene is a member of serine/threonine protein kinase family. This kinase preferentially activates other kinases involved in the MAP kinase signaling pathway. This kinase has been shown to directly phosphorylate and activate Ikappa B kinases, and thus plays a role in NF-kappa B signaling pathway. This kinase has also been found to bind and activate protein kinase C-related kinase 2, which suggests its involvement in a regulated signaling process. [provided by RefSeq]
Sequence: HMKSLKILLVINGSTQATNLEPLPSLEDLDNTVFGAERKKRLSIIGPTSRDRSSPPPGYIPDELHQVARNGSFTSINSEGEFIPESMEQM
Especificaciones
Especificaciones
MAP3K2 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
35.53kDa