missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAP3K11 Partial ORF (AAH64543, 499 a.a. - 599 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004296-Q01.25ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene is a member of the serine/threonine kinase family. This kinase contains a SH3 domain and a leucine zipper-basic motif. This kinase preferentially activates MAPK8/JNK kinase, and functions as a positive regulator of JNK signaling pathway. This kinase can directly phosphorylate, and activates IkappaB kinase alpha and beta, and is found to be involved in the transcription activity of NF-kappaB mediated by Rho family GTPases and CDC42. [provided by RefSeq]
Sequence: KHRITVQASPGLDRRRNVFEVGPGDSPTFPRFRAIQLEPAEPGQAWGRQSPRRLEDSSNGERRACWAWGPSSPKPGEAQNGRRRSRMDEATWYLDSDDSSPEspecificaciones
AAH64543 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KHRITVQASPGLDRRRNVFEVGPGDSPTFPRFRAIQLEPAEPGQAWGRQSPRRLEDSSNGERRACWAWGPSSPKPGEAQNGRRRSRMDEATWYLDSDDSSP | |
RUO | |
MAP3K11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4296 | |
MAP3K11 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC17114/MLK-3/MLK3/PTK1/SPRK | |
MAP3K11 | |
Recombinant | |
wheat germ expression system |