missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAML1 Partial ORF (NP_055572, 655 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00009794-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16171286

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 07-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This protein is the human homolog of mastermind, a Drosophila protein that plays a role in the Notch signaling pathway involved in cell-fate determination. There is in vitro evidence that the human homolog forms a complex with the intracellular portion of human Notch receptors and can increase expression of a Notch-induced gene. This evidence supports its proposed function as a transcriptional co-activator in the Notch signaling pathway. [provided by RefSeq]

Sequence: AEQEKQQFQRHLTRPPPQYQDPTQGSFPQQVGQFTGSSAAVPGMNTLGPSNSSCPRVFPQAGNLMPMGPGHASVSSLPTNSGQQDRGVAQFPGSQNMPQS
Especificaciones

Especificaciones

NP_055572
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AEQEKQQFQRHLTRPPPQYQDPTQGSFPQQVGQFTGSSAAVPGMNTLGPSNSSCPRVFPQAGNLMPMGPGHASVSSLPTNSGQQDRGVAQFPGSQNMPQS
RUO
MAML1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9794
MAML1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0200/Mam-1/Mam1
MAML1
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales