Learn More
Abnova™ Human MAGEB6 Full-length ORF (NP_775794.2, 1 a.a. - 407 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marque: Abnova™ H00158809-P01.10ug
Informations supplémentaires : Peso : 0.00010kg
Description
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is expressed in testis, and in a significant fraction of tumors of various histological types. The MAGEB genes are clustered on chromosome Xp22-p21. [provided by RefSeq]
Sequence: MPRGHKSKLRTCEKRQETNGQPQGLTGPQATAEKQEESHSSSSSSRACLGDCRRSSDASIPQESQGVSPTGSPDAVVSYSKSDVAANGQDEKSPSTSRDASVPQESQGASPTGSPDAGVSGSKYDVAANGQDEKSPSTSHDVSVPQESQGASPTGSPDAGVSGSKYDVAAEGEDEESVSASQKAIIFKRLSKDAVKKKACTLAQFLQKKFEKKESILKADMLKCVRREYKPYFPQILNRTSQHLVVAFGVELKEMDSSGESYTLVSKLGLPSEGILSGDNALPKSGLLMSLLVVIFMNGNCATEEEVWEFLGLLGIYDGILHSIYGDARKIITEDLVQDKYVVYRQVCNSDPPCYEFLWGPRAYAETTKMRVLRVLADSSNTSPGLYPHLYEDALIDEVERALRLRASpécification
NP_775794.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ40242/MAGE-B6/MAGEB6A | |
MAGEB6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
158809 | |
MAGEB6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPRGHKSKLRTCEKRQETNGQPQGLTGPQATAEKQEESHSSSSSSRACLGDCRRSSDASIPQESQGVSPTGSPDAVVSYSKSDVAANGQDEKSPSTSRDASVPQESQGASPTGSPDAGVSGSKYDVAANGQDEKSPSTSHDVSVPQESQGASPTGSPDAGVSGSKYDVAAEGEDEESVSASQKAIIFKRLSKDAVKKKACTLAQFLQKKFEKKESILKADMLKCVRREYKPYFPQILNRTSQHLVVAFGVELKEMDSSGESYTLVSKLGLPSEGILSGDNALPKSGLLMSLLVVIFMNGNCATEEEVWEFLGLLGIYDGILHSIYGDARKIITEDLVQDKYVVYRQVCNSDPPCYEFLWGPRAYAETTKMRVLRVLADSSNTSPGLYPHLYEDALIDEVERALRLRA | |
RUO | |
MAGEB6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |