Learn More
Abnova™ Human LYST Partial ORF (NP_000072.2, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001130-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is thought to encode a protein that regulates intracellular protein trafficking to and from the lysosome. Mutations in this gene are associated with Chediak-Higashi syndrome, a rare lysosomal storage disorder. Alternatively spliced transcript variants have been described but their full-length nature has not been determined. [provided by RefSeq]
Sequence: MSTDSNSLAREFLTDVNRLCNAVVQRVEAREEEEEETHMATLGQYLVHGRGFLLLTKLNSIIDQALTCREELLTLLLSLLPLVWKIPVQEEKATDFNLPLEspecificaciones
NP_000072.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSTDSNSLAREFLTDVNRLCNAVVQRVEAREEEEEETHMATLGQYLVHGRGFLLLTKLNSIIDQALTCREELLTLLLSLLPLVWKIPVQEEKATDFNLPL | |
RUO | |
LYST | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1130 | |
LYST (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CHS/CHS1 | |
LYST | |
Recombinant | |
wheat germ expression system |