missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LYK5 Partial ORF (NP_699166.2, 251 a.a. - 346 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00092335-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16171207

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 07-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene contains a STE20-like kinase domain, but lacks several residues that are critical for catalytic activity, so it is termed a 'pseudokinase'. The protein forms a heterotrimeric complex with serine/threonine kinase 11 (STK11, also known as LKB1) and the scaffolding protein calcium binding protein 39 (CAB39, also known as MO25). The protein activates STK11 leading to the phosphorylation of both proteins and excluding STK11 from the nucleus. The protein is necessary for STK11-induced G1 cell cycle arrest. A mutation in this gene has been shown to result in polyhydramnios, megalencephaly, and symptomatic epilepsy (PMSE) syndrome. Multiple transcript variants encoding different isoforms have been found for this gene. Additional transcript variants have been described but their full-length nature is not known. [provided by RefSeq]

Sequence: LLEKLNGTVPCLLDTSTIPAEELTMSPSRSVANSGLSDSLTTSTPRPSNGDSPSHPYHRTFSPHFHHFVEQCLQRNPDARYPCWPGPGLRESRGCS
Especificaciones

Especificaciones

NP_699166.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LLEKLNGTVPCLLDTSTIPAEELTMSPSRSVANSGLSDSLTTSTPRPSNGDSPSHPYHRTFSPHFHHFVEQCLQRNPDARYPCWPGPGLRESRGCS
RUO
STRADA
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
92335
LYK5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ90524/LYK5/NY-BR-96/PMSE/STRAD/Stlk
STRADA
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales