Learn More
Abnova™ Human LRSAM1 Full-length ORF (NP_001005373.1, 1 a.a. - 723 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00090678-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
LRSAM1 is a multifunctional RING finger protein that selectively regulates cell adhesion molecules, has ubiquitin ligase activity, and plays a role in receptor endocytosis and viral budding (Li et al., 2003 [PubMed 14635195]; Amit et al., 2004 [PubMed 15256501]).[supplied by OMIM]
Sequence: MPLFFRKRKPSEEARKRLEYQMCLAKEAGADDILDISKCELSEIPFGAFATCKVLQKKVLIVHTNHLTSLLPKSCSLLSLATIKVLDLHDNQLTALPDDLGQLTALQVLNVERNQLMQLPRSIGNLTQLQTLNVKDNKLKELPDTVGELRSLRTLNISGNEIQRLPQMLAHVRTLEMLSLDASAMVYPPREVCGAGTAAILQFLCKESGLEYYPPSQYLLPILEQDGIENSRDSPDGPTDRFSREELEWQNRFSDYEKRKEQKMLEKLEFERRLELGQREHTQLLQQSSSQKDEILQTVKEEQSRLEQGLSEHQRHLNAERQRLQEQLKQTEQNISSRIQKLLQDNQRQKKSSEILKSLENERIRMEQLMSITQEETESLRRRDVASAMQQMLTESCKNRLIQMAYESQRQNLVQQACSSMAEMDERFQQILSWQQMDQNKAISQILQESAMQKAAFEALQVKKDLMHRQIRSQIKLIETELLQLTQLELKRKSLDTESLQEMISEQRWALSSLLQQLLKEKQQREEELREILTELEAKSETRQENYWLIQYQRLLNQKPLSLKLQEEGMERQLVALLEELSAEHYLPIFAHHRLSLDLLSQMSPGDLAKVGVSEAGLQHEILRRVQELLDAARIQPELKPPMGEVVTPTAPQEPPESVRPSAPPAELEVQASECVVCLEREAQMIFLNCGHVCCCQQCCQPLRTCPLCRQDIAQRLRIYHSSEspecificaciones
NP_001005373.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ31641/RIFLE/TAL | |
LRSAM1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
90678 | |
LRSAM1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPLFFRKRKPSEEARKRLEYQMCLAKEAGADDILDISKCELSEIPFGAFATCKVLQKKVLIVHTNHLTSLLPKSCSLLSLATIKVLDLHDNQLTALPDDLGQLTALQVLNVERNQLMQLPRSIGNLTQLQTLNVKDNKLKELPDTVGELRSLRTLNISGNEIQRLPQMLAHVRTLEMLSLDASAMVYPPREVCGAGTAAILQFLCKESGLEYYPPSQYLLPILEQDGIENSRDSPDGPTDRFSREELEWQNRFSDYEKRKEQKMLEKLEFERRLELGQREHTQLLQQSSSQKDEILQTVKEEQSRLEQGLSEHQRHLNAERQRLQEQLKQTEQNISSRIQKLLQDNQRQKKSSEILKSLENERIRMEQLMSITQEETESLRRRDVASAMQQMLTESCKNRLIQMAYESQRQNLVQQACSSMAEMDERFQQILSWQQMDQNKAISQILQESAMQKAAFEALQVKKDLMHRQIRSQIKLIETELLQLTQLELKRKSLDTESLQEMISEQRWALSSLLQQLLKEKQQREEELREILTELEAKSETRQENYWLIQYQRLLNQKPLSLKLQEEGMERQLVALLEELSAEHYLPIFAHHRLSLDLLSQMSPGDLAKVGVSEAGLQHEILRRVQELLDAARIQPELKPPMGEVVTPTAPQEPPESVRPSAPPAELEVQASECVVCLEREAQMIFLNCGHVCCCQQCCQPLRTCPLCRQDIAQRLRIYHSS | |
RUO | |
LRSAM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |