missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LRSAM1 Full-length ORF (NP_001005373.1, 1 a.a. - 723 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00090678-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16196523

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 06-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

LRSAM1 is a multifunctional RING finger protein that selectively regulates cell adhesion molecules, has ubiquitin ligase activity, and plays a role in receptor endocytosis and viral budding (Li et al., 2003 [PubMed 14635195]; Amit et al., 2004 [PubMed 15256501]).[supplied by OMIM]

Sequence: MPLFFRKRKPSEEARKRLEYQMCLAKEAGADDILDISKCELSEIPFGAFATCKVLQKKVLIVHTNHLTSLLPKSCSLLSLATIKVLDLHDNQLTALPDDLGQLTALQVLNVERNQLMQLPRSIGNLTQLQTLNVKDNKLKELPDTVGELRSLRTLNISGNEIQRLPQMLAHVRTLEMLSLDASAMVYPPREVCGAGTAAILQFLCKESGLEYYPPSQYLLPILEQDGIENSRDSPDGPTDRFSREELEWQNRFSDYEKRKEQKMLEKLEFERRLELGQREHTQLLQQSSSQKDEILQTVKEEQSRLEQGLSEHQRHLNAERQRLQEQLKQTEQNISSRIQKLLQDNQRQKKSSEILKSLENERIRMEQLMSITQEETESLRRRDVASAMQQMLTESCKNRLIQMAYESQRQNLVQQACSSMAEMDERFQQILSWQQMDQNKAISQILQESAMQKAAFEALQVKKDLMHRQIRSQIKLIETELLQLTQLELKRKSLDTESLQEMISEQRWALSSLLQQLLKEKQQREEELREILTELEAKSETRQENYWLIQYQRLLNQKPLSLKLQEEGMERQLVALLEELSAEHYLPIFAHHRLSLDLLSQMSPGDLAKVGVSEAGLQHEILRRVQELLDAARIQPELKPPMGEVVTPTAPQEPPESVRPSAPPAELEVQASECVVCLEREAQMIFLNCGHVCCCQQCCQPLRTCPLCRQDIAQRLRIYHSS
Especificaciones

Especificaciones

NP_001005373.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
110kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ31641/RIFLE/TAL
LRSAM1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
90678
LRSAM1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPLFFRKRKPSEEARKRLEYQMCLAKEAGADDILDISKCELSEIPFGAFATCKVLQKKVLIVHTNHLTSLLPKSCSLLSLATIKVLDLHDNQLTALPDDLGQLTALQVLNVERNQLMQLPRSIGNLTQLQTLNVKDNKLKELPDTVGELRSLRTLNISGNEIQRLPQMLAHVRTLEMLSLDASAMVYPPREVCGAGTAAILQFLCKESGLEYYPPSQYLLPILEQDGIENSRDSPDGPTDRFSREELEWQNRFSDYEKRKEQKMLEKLEFERRLELGQREHTQLLQQSSSQKDEILQTVKEEQSRLEQGLSEHQRHLNAERQRLQEQLKQTEQNISSRIQKLLQDNQRQKKSSEILKSLENERIRMEQLMSITQEETESLRRRDVASAMQQMLTESCKNRLIQMAYESQRQNLVQQACSSMAEMDERFQQILSWQQMDQNKAISQILQESAMQKAAFEALQVKKDLMHRQIRSQIKLIETELLQLTQLELKRKSLDTESLQEMISEQRWALSSLLQQLLKEKQQREEELREILTELEAKSETRQENYWLIQYQRLLNQKPLSLKLQEEGMERQLVALLEELSAEHYLPIFAHHRLSLDLLSQMSPGDLAKVGVSEAGLQHEILRRVQELLDAARIQPELKPPMGEVVTPTAPQEPPESVRPSAPPAELEVQASECVVCLEREAQMIFLNCGHVCCCQQCCQPLRTCPLCRQDIAQRLRIYHSS
RUO
LRSAM1
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales