Learn More
Abnova™ Human LRP8 Partial ORF (NP_004622, 83 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007804-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes an apolipoprotein E receptor, a member of the low density lipoprotein receptor (LDLR) family. Apolipoprotein E is a small lipophilic plasma protein and a component of lipoproteins such as chylomicron remnants, very low density lipoprotein (VLDL), and high density lipoprotein (HDL). The apolipoprotein E receptor is involved in cellular recognition and internalization of these lipoproteins. Alternative splicing generates multiple transcript variants encoding distinct isoforms for this gene. [provided by RefSeq]
Sequence: KKTCADSDFTCDNGHCIHERWKCDGEEECPDGSDESEATCTKQVCPAEKLSCGPTSHKCVPASWRCDGEKDCEGGADEAGCATLCAPHEspecificaciones
NP_004622 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KKTCADSDFTCDNGHCIHERWKCDGEEECPDGSDESEATCTKQVCPAEKLSCGPTSHKCVPASWRCDGEKDCEGGADEAGCATLCAPH | |
RUO | |
LRP8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7804 | |
LRP8 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APOER2/HSZ75190/MCI1 | |
LRP8 | |
Recombinant | |
wheat germ expression system |