missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LPIN1 Partial ORF (NP_663731.1, 792 a.a. - 890 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00023175-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16164456

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 13-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene represents a candidate gene for human lipodystrophy, characterized by loss of body fat, fatty liver, hypertriglyceridemia, and insulin resistance. Mouse studies suggest that this gene functions during normal adipose tissue development and may also play a role in human triglyceride metabolism. [provided by RefSeq]

Sequence: EPFYAAFGNRPADVYSYKQVGVSLNRIFTVNPKGELVQEHAKTNISSYVRLCEVVDHVFPLLKRSHSSDFPCSDTFSNFTFWREPLPPFENQDIHSASA
Especificaciones

Especificaciones

NP_663731.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EPFYAAFGNRPADVYSYKQVGVSLNRIFTVNPKGELVQEHAKTNISSYVRLCEVVDHVFPLLKRSHSSDFPCSDTFSNFTFWREPLPPFENQDIHSASA
RUO
LPIN1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
23175
LPIN1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp781P1796/KIAA0188/PAP1
LPIN1
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales