missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LOC375748 Full-length ORF (AAH35183.1, 1 a.a. - 523 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00375748-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MQKERELQETIYFKNTWPACMFLIVAPLSVLYNWKDELDTWGYFRVTVLHGNRKDNELIRVKQRKCEIALTTYETLRLCLDELNSLEWSAVIVDEAHRIKNPKARVTEVMKALKCNVRIGLTGTILQNNMKELWCVMDWAVPGLLGSGTYFKKQFSDPVEHGQRHTATKRELATGRKAMQRLAKKMSGWFLRRTKTLIKDQLPKKEDRMVYCSLTDFQKAVYQTVLETEDVTLILQSSEPCTCRSGQKRRNCCYKTNSHGETVKTLYLSYLTVLQKVANHVALLQAASTSKQQETLIKRICDQVFSRFPDFVQKSKDAAFETLSDPKYSGKMKVLQQLLNHCRKNRDKVLLFSFSTKLLDVLQQYCMASGLDYRRLDGSTKSEERLKIVKEFNSTQDVNICLVSTMAGGLGLNFVGANVVVLFDPTWNPANDLQAIDRAYRIGQCRDVKVLRLISLGTVEEIMYLRQIYKQQLHCVVVGSENAKRYFEAVQGSKEHQGELFGIHNLFKFRSQGSCLTKDILEVEspecificaciones
AAH35183.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
86.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ37706/MGC30192/MGC43364/RAD26L/SR278 | |
C9orf102 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
375748 | |
LOC375748 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQKERELQETIYFKNTWPACMFLIVAPLSVLYNWKDELDTWGYFRVTVLHGNRKDNELIRVKQRKCEIALTTYETLRLCLDELNSLEWSAVIVDEAHRIKNPKARVTEVMKALKCNVRIGLTGTILQNNMKELWCVMDWAVPGLLGSGTYFKKQFSDPVEHGQRHTATKRELATGRKAMQRLAKKMSGWFLRRTKTLIKDQLPKKEDRMVYCSLTDFQKAVYQTVLETEDVTLILQSSEPCTCRSGQKRRNCCYKTNSHGETVKTLYLSYLTVLQKVANHVALLQAASTSKQQETLIKRICDQVFSRFPDFVQKSKDAAFETLSDPKYSGKMKVLQQLLNHCRKNRDKVLLFSFSTKLLDVLQQYCMASGLDYRRLDGSTKSEERLKIVKEFNSTQDVNICLVSTMAGGLGLNFVGANVVVLFDPTWNPANDLQAIDRAYRIGQCRDVKVLRLISLGTVEEIMYLRQIYKQQLHCVVVGSENAKRYFEAVQGSKEHQGELFGIHNLFKFRSQGSCLTKDILEV | |
RUO | |
C9orf102 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |