Learn More
Abnova™ Human LOC150577 Full-length ORF (XP_379114.1, 1 a.a. - 92 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00150577-P01.25ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16168623
  • 508.00€ / 25 microgramos
Fecha estimada de envío: 13-06-2024
para ver el stock
Descripción
Descripción
Sequence: MVHSRKWGESGQRCPGGLCLHHLERHRPFSLHSLFRKEAYSQMNAPRLRDKANPWDAEGTWITSARERSPNSASRRPEVWPPVDTQLRDEQQ
Especificaciones
Especificaciones
XP_379114.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LOC150577
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
150577
LOC150577 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVHSRKWGESGQRCPGGLCLHHLERHRPFSLHSLFRKEAYSQMNAPRLRDKANPWDAEGTWITSARERSPNSASRRPEVWPPVDTQLRDEQQ
RUO
LOC150577
Recombinant
wheat germ expression system